Learn More
Memmert™ Entry Port 14 mm for UF160plus
Shop All Memmert Products
Click to view available options
Diameter (Metric):
14 mm
Unit Size:
Each
Description
- Entry port with 14 mm diameter
- Flap closure
- Custom position TBC in the back
Specifications
Specifications
| For Use With (Equipment) | UF160plus Oven |
| Type | Entry Port |
| Diameter (Metric) | 14 mm |