Learn More
Memmert™ Entry Port 14 mm, Flap Closure, Custom Position Tbc In The Back For Ipp55
Product Code. 15387307 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
IPP55 Peltier-cooled Incubator
Unit Size:
Each
Product Code. 15387307
Brand: Memmert™ IPP55D6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Optional entry port for cooled incubator. Memmert™ Entry Port, Pelitier-Cooled Incubator can be placed in special positions at the back (specify).
  • Can be closed by flap
  • In special positions on the back
  • 14 mm clear diameter
Specifications
For Use With (Equipment) IPP55 Peltier-cooled Incubator
Type Entry Port
Length (Metric) 14 mm