missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENTPD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115181
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ENTPD5.
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
Specifications
| ENTPD5 | |
| Polyclonal | |
| Unconjugated | |
| ENTPD5 | |
| AI196558; AI987697; CD39 antigen-like 4; CD39L4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; Entpd5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; mNTPase; NTPDase 5; NTPDase5; NTPDase-5; Nucleoside diphosphatase; PCPH; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 957 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O75356 | |
| ENTPD5 | |
| A synthesized peptide derived from human ENTPD5(Accession O75356), corresponding to amino acid residues S85-L135. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction