missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENTPD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112412
This item is not returnable.
View return policy
Description
ENTPD5 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
Specifications
| ENTPD5 | |
| Polyclonal | |
| Unconjugated | |
| ENTPD5 | |
| AI196558; AI987697; CD39 antigen-like 4; CD39L4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; Entpd5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; mNTPase; NTPDase 5; NTPDase5; NTPDase-5; Nucleoside diphosphatase; PCPH; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12499, 957 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75356, Q9WUZ9 | |
| ENTPD5 | |
| Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 5 protein (149-428AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction