missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENTPD5 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA547716
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.
Specifications
| ENTPD5 | |
| Polyclonal | |
| Unconjugated | |
| ENTPD5 | |
| AI196558; AI987697; CD39 antigen-like 4; CD39L4; CD39-like 4; Ectonucleoside triphosphate diphosphohydrolase 5; Entpd5; ER-UDPase; GDPase ENTPD5; Guanosine-diphosphatase ENTPD5; mNTPase; NTPDase 5; NTPDase5; NTPDase-5; Nucleoside diphosphatase; PCPH; Pcph proto-oncogene protein; proto-oncogene CPH; UDPase ENTPD5; Uridine-diphosphatase ENTPD5 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 957 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O75356 | |
| ENTPD5 | |
| CHO-derived recombinant human CD39L4/ENTPD5 Val21-His428. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction