missing translation for 'onlineSavingsMsg'
Learn More

ENTPD4 Antibody, Novus Biologicals™

Product Code. 18368948 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368948 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368948 Supplier Novus Biologicals Supplier No. H00009583B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ENTPD4 Polyclonal antibody specifically detects ENTPD4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH34477.1
Gene Alias ectonucleoside triphosphate diphosphohydrolase 4, KIAA0392NTPDase 4, LALP70EC 3.6.1.6, LAP70guanosine-diphosphatase like protein, LYSAL1uridine-diphosphatase, lysosomal apyrase-like 1, Lysosomal apyrase-like protein of 70 kDa, NTPDase-4, UDPaseapyrase, lysosomal, Uridine-diphosphatase
Host Species Mouse
Immunogen ENTPD4 (AAH34477.1, 1 a.a. - 571 a.a.) full-length human protein. MGRIGISCLFPASWHFSISPVGCPRILNTNLRQIMVISVLAAAVSLLYFSVVIIRNKYGRLTRDKKFQRYLARVTDIEATDTNNPNVNYGIVVDCGSSGSRVFVYCWPRHNGNPHDLLDIRQMRDKNRKPVVMKIKPGISEFATSPEKVSDYISPLLNFAAEHVPRAKHKETPLYILCTAGMRILPESQQKAILEDLLTDIPVHFDFLFSDSHAEVISGKQEGVYAWIGINFVLGRFEHIEDDDEAVVEVNIPGSESSEAIVRKRTAGILDMGGVSTQIAYEVPKTEEVAKNLLAEFNLGCDVHQTEHVYRVYVATFLGFGGNAARQRYEDRIFANTIQKNRLLGKQTGLTPDMPYLDPCLPLDIKDEIQQNGQTIYLRGTGDFDLCRETIQPFMNKTNETQTSLNGVYQPPIHFQNSEFYGFSEFYYCTEDVLRMGGDYNAAKFTKAAKDYCATKWSILRERFDRGLYASHADLHRLKYQCFKSAWMFEVFHRGFSFPVNYKSLKTALQVYDKEVQWTLGAILYRTRFLPLRKIGMLIIPCSGGCPKLCCAVWQPPPYGATKHLQCAAWN
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9583
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.