missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENTPD3 Polyclonal Antibody

Description
In direct ELISAs, approximately 30% cross-reactivity with recombinant human CD39L3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
T-cadherin was initially identified as cadherin-type cell adhesion molecule expressed in various neuronal populations in a temporally and spatially restricted pattern during axon growth. T-cadherin is an atypical member of the cadherin family because it does not possess the typical transmembrane and cytoplasmic domains but is instead anchored to the plasma membrane by glycosylphosphatidylinositol (GPI) linkage. T-cadherin may play a role in malignant tumor development as loss of the chromosome locus containing the T-cadherin gene correlates with the development of a variety of cancers. Recently it has been shown that T-cadherin can act as a receptor for hexameric and high-molecular weight forms of adiponectin, suggesting that T-cadherin may also play a role in metabolic regulation.
Specifications
Specifications
| Antigen | ENTPD3 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | ENTPD3 |
| Gene Accession No. | 0 |
| Gene Alias | CD39 antigen-like 3; CD39L3; CD39-like 3; ecto-apyrase 3; Ecto-ATP diphosphohydrolase 3; ecto-ATPase 3; ecto-ATPDase 3; Ectonucleoside triphosphate diphosphohydrolase 3; ENTPD3; HB6; NTPDase 3; NTPDase3; NTPDase-3 |
| Gene Symbols | ENTPD3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?