missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENTPD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13474419
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13474419 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13474419 Supplier Invitrogen™ Supplier No. PA532750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Panda, Dog, Horse (94%) Marmoset, Mouse, Rat, Hamster, Elephant, Bovine (88%).

The protein encoded by this gene is the type 2 enzyme of the ecto-nucleoside triphosphate diphosphohydrolase family (E-NTPDase). E-NTPDases are a family of ecto-nucleosidases that hydrolyze 5'-triphosphates. This ecto-ATPase is an integral membrane protein. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENTPD2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ENTPD2
Gene Accession No. Q9Y5L3
Gene Alias CD39 antigen-like 1; CD39L1; Ecto-ATP diphosphohydrolase 2; ecto-ATPase 2; ecto-ATPDase 2; Ectonucleoside triphosphate diphosphohydrolase 2; ENTPD2; NTPDase 2; NTPDase2; NTPDase-2; testicular ecto-ATPase
Gene Symbols ENTPD2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from internal region of human ENTPD2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 954
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.