missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entinostat, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, respectively.
Specifications
Specifications
| Chemical Name or Material | Entinostat |
| Physical Form | Solid |
| CAS | 209783-80-2 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H20N4O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | MS-275 SNDX-275 |
| Solubility Information | DMSO : 50 mg/mL (132.83 mM; ultrasonic and warming and heat to 60°C) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction