missing translation for 'onlineSavingsMsg'
Learn More

Entinostat, MedChemExpress

Product Code. 16421036
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16421036

Brand: Medchem Express HY12163

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, respectively.

Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, respectively.

Specifications

Chemical Name or Material Entinostat
Physical Form Solid
CAS 209783-80-2
Color Off-White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C21H20N4O3
For Use With (Application) Cancer-programmed cell death
Synonym MS-275 SNDX-275
Solubility Information DMSO : 50 mg/mL (132.83 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NCC1=CC=C(C=C1)C(NC2=CC=CC=C2N)=O)OCC3=CC=CN=C3
Molecular Weight (g/mol) 376.41
Formula Weight 376.41
Percent Purity 98.07%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.