missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENTH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112387
This item is not returnable.
View return policy
Description
ENTH Polyclonal Antibody for Western Blot, IHC (P), ELISA
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.
Specifications
| ENTH | |
| Polyclonal | |
| Unconjugated | |
| CLINT1 | |
| AI642036; AW537616; C530049I24Rik; clathrin interacting protein localized in the trans-Golgi region; clathrin interactor 1; Clathrin interactor 1-like protein; Clathrin-interacting protein localized in the trans-Golgi region; Clint; Clint1; ENTH; ENTH domain containing protein; enthoprotin; Epn4; EPNR; epsin 4; epsin-4; epsinR; Epsin-related protein; F630206G17Rik; KIAA0171; mKIAA0171 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 216705, 360515, 9685 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q14677, Q99KN9 | |
| CLINT1 | |
| Recombinant Human Clathrin interactor 1 protein (1-625AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction