missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Enterovirus D68 VP3 (Strain: 37-99) Recombinant Rabbit Monoclonal Antibody (HL3202)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556697
This item is not returnable.
View return policy
Description
Enterovirus D68 VP3 (Strain: 37-99) Recombinant Monoclonal Antibody for Western Blot
Enterovirus D68 (EV-D68) has been recognized as an emerging pathogen after recent worldwide outbreaks of serious respiratory and neurological disease. Its capside proteins are VP1-VP4.
Specifications
| Enterovirus D68 VP3 (Strain: 37-99) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3202 | |
| Unconjugated | |
| Enterovirus | |
| Recombinant fragment of Enterovirus D68 VP3. (Strain# 37-99). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction