missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Enterovirus 71 VP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111998
This item is not returnable.
View return policy
Description
Enterovirus 71 VP3 Polyclonal Antibody for Western Blot, ICC/IF
Enterovirus 71 (EV71) causes hand-foot-and-mouth disease (HFMD), for which there are currently no therapeutics. EV71 virions have icosahedral symmetry and a diameter of approximately 30 nm, comprising 60 copies of the four protein subunits VP1 to VP4. Each of subunits VP1 to VP3 adopts a beta-barrel configuration common to many viruses.
Specifications
| Enterovirus 71 VP3 | |
| Polyclonal | |
| Unconjugated | |
| Enterovirus | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of Enterovirus 71 VP3 (EV71 virus (strain USA/BrCr/1970)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.2 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction