missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Enterovirus 71 3CD Polyclonal antibody specifically detects Enterovirus 71 3CD in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Enterovirus 71 3CD |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | EV71 3CD, Protein 3CD |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of Enterovirus 71 3CD (EV71 virus (strain USA/BrCr/1970)). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?