missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Enterostatin(human,mouse,rat)

Product Code. 16422829
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422829 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422829 Supplier MedChemExpress Supplier No. HYP1067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enterostatin, human, mouse, rat is a pentapeptide that reduces fat intake.

Specifications

Purity 99.87%
CAS 117830-79-2
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Metabolism-sugar/lipid metabolism
Molecular Weight (g/mol) 496.56 g/mol
Product Type Natural Products
Quantity 1 mg
Shipping Condition Room temperature
Solubility DMSO : 125 mg/mL (251.73 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.