missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterobacter aerogenes Mouse anti-Bacteria, Clone: 57/82, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Enterobacter aerogenes Monoclonal specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Enterobacter aerogenes |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 57/82 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Host Species | Mouse |
| Immunogen | A crude sonicate of Enterobacter aerogenes |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?