missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enterobacter aerogenes Mouse anti-Bacteria, Clone: 57/82, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | Enterobacter aerogenes |
|---|---|
| Clone | 57/82 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunofluorescence |
Description
Enterobacter aerogenes Monoclonal specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Enterobacter aerogenes | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| A crude sonicate of Enterobacter aerogenes | |
| Primary | |
| Store at 4°C. |
| 57/82 | |
| ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title