missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Entecavir Monohydrate, Mikromol™
Description
Entecavir Monohydrate
Specifications
Specifications
| Chemical Name or Material | Entecavir Monohydrate |
| CAS | 209216-23-9 |
| Analyte or Component Names | Entecavir Monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H15 N5 O3 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, hydrate (1:1), 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, monohydrate (9CI), Entecavir monohydrate |
| InChI Formula | InChI=1S/C12H15N5O3.H2O/c1-5-6(3-18)8(19)2-7(5)17-4-14-9-10(17)15-12(13)16-11(9)20;/h4,6-8,18-19H,1-3H2,(H3,13,15,16,20);1H2/t6-,7-,8-;/m0./s1 |
| SMILES | O.NC1=Nc2c(ncn2[C@H]3C[C@H](O)[C@@H](CO)C3=C)C(=O)N1 |
| IUPAC Name | 2-amino-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylidenecyclopentyl]-1H-purin-6-one;hydrate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?