missing translation for 'onlineSavingsMsg'
Learn More

Entecavir monohydrate, MedChemExpress

Product Code. 16435778
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435778

Brand: Medchem Express HY13623A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Entecavir monohydrate (BMS200475 monohydrate; SQ34676 monohydrate) is a potent and selective inhibitor of HBV, with an EC50 of 3.75 nM in HepG2 cell.

Entecavir monohydrate (BMS200475 monohydrate

Specifications

Chemical Name or Material Entecavir monohydrate
CAS 209216-23-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C12H17N5O4
Quantity 10 mg
Synonym BMS200475 monohydrateSQ34676 monohydrate
Solubility Information DMSO : ≥ 50 mg/mL (169.33 mM) ∣H2O : 2.8 mg/mL (9.48 mM; Need ultrasonic and warming)
SMILES O=C1NC(N)=NC2=C1N=CN2[C@@H]3C([C@H](CO)[C@@H](O)C3)=C.O
Molecular Weight (g/mol) 295.29
Formula Weight 295.29
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.