missing translation for 'onlineSavingsMsg'
Learn More

Entasobulin, MedChemExpress

Product Code. 16489717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489717 5 mg 5mg
16487188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489717 Supplier Medchem Express Supplier No. HY16777
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Entasobulin is a β-tubulin polymerization inhibitor with potential anticancer activity.

Entasobulin is a β-tubulin polymerization inhibitor with potential anticancer activity.

Specifications

Chemical Name or Material Entasobulin
CAS 501921-61-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H18ClN3O2
Quantity 5 mg
Solubility Information DMSO : 155 mg/mL (352.36 mM; Need ultrasonic)
SMILES O=C(C(NC1=CC(C=CC=N2)=C2C=C1)=O)C3=CN(C4=CC=CC=C43)CC5=CC=C(Cl)C=C5
Molecular Weight (g/mol) 439.89
Formula Weight 439.89
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.