missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Entacapone

Product Code. 17291202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17291202 250 mg 250mg
17281202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17291202 Supplier Thermo Scientific Chemicals Supplier No. 466222500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130929-57-6
Molecular Formula C14H15N3O5

Specifications

Quantity 250 mg
Formula Weight 305.29
InChI Key JRURYQJSLYLRLN-BJMVGYQFSA-N
SMILES CCN(CC)C(=O)C(=C\C1=CC(O)=C(O)C(=C1)[N+]([O-])=O)\C#N
IUPAC Name (2E)-2-cyano-3-(3,4-dihydroxy-5-nitrophenyl)-N,N-diethylprop-2-enamide
Molecular Weight (g/mol) 305.29
Chemical Name or Material Entacapone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.