missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENT2 Recombinant Rabbit Monoclonal Antibody (24GB2370)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557258
This item is not returnable.
View return policy
Description
ENT2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SLC29A2 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A2 is a transmembrane glycoprotein that mediates the cellular uptake of nucleosides from the surrounding medium. As a nucleoside transporter, SLC29A2 plays an important role in the uptake of nucleoside-based anti-cancer drugs; polymorphisms of point mutations in the gene encoding this protein may affect the efficacy of these drugs.
Specifications
| ENT2 | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O54699, Q14542, Q61672 | |
| SLC29A2 | |
| A synthesized peptide derived from human ENT2 (aa 1-29). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2370 | |
| Unconjugated | |
| SLC29A2 | |
| 36 kDa hydrophobic nucleolar protein; 36 kDa nucleolar protein HNP36; delayed early response gene 12; delayed-early response protein 12; DER12; ENT2; equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; equilibrative nucleoside transporter 2; HNP36; hydrophobic nucleolar protein, 36 kDa; hydrophobic nucleolar protein, 36kD; NBMPR-insensitive nucleoside transporter ei; NBMPR-insensitive nucleoside transporter ei 2A; nucleoside transporter, ei-type; rbENT2A; Slc29a2; solute carrier family 29 (equilibrative nucleoside transporter), member 2; solute carrier family 29 (nucleoside transporters), member 2; solute carrier family 29 member 2; solute carrier family 29, member 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13340, 3177, 65194 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction