missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553746
This item is not returnable.
View return policy
Description
Immunogen sequence: KFARYYLANK SSQAQAQELE TKAELLQSDE NGIPSSPQKV ALTLDLDLEK EPESEPDEPQ KPGKPSVFTV FQK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.
SLC29A2 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A2 is a transmembrane glycoprotein that mediates the cellular uptake of nucleosides from the surrounding medium. As a nucleoside transporter, SLC29A2 plays an important role in the uptake of nucleoside-based anti-cancer drugs; polymorphisms of point mutations in the gene encoding this protein may affect the efficacy of these drugs.
Specifications
| ENT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC29A2 | |
| 36 kDa hydrophobic nucleolar protein; 36 kDa nucleolar protein HNP36; delayed early response gene 12; delayed-early response protein 12; DER12; ENT2; equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; equilibrative nucleoside transporter 2; HNP36; hydrophobic nucleolar protein, 36 kDa; hydrophobic nucleolar protein, 36kD; NBMPR-insensitive nucleoside transporter ei; NBMPR-insensitive nucleoside transporter ei 2A; nucleoside transporter, ei-type; rbENT2A; Slc29a2; solute carrier family 29 (equilibrative nucleoside transporter), member 2; solute carrier family 29 (nucleoside transporters), member 2; solute carrier family 29 member 2; solute carrier family 29, member 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3177 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14542 | |
| SLC29A2 | |
| Recombinant protein corresponding to Human SLC29A2. Recombinant protein control fragment (Product #RP-91298). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction