missing translation for 'onlineSavingsMsg'
Learn More

ent-Lamivudine, TRC

Product Code. 30475354
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475354 1 mg 1mg
30470409 10 mg 10mg
30468328 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475354 Supplier Toronto Research Chemicals Supplier No. TRCL1725051MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134680-32-3
Molecular Formula C8H11N3O3S
Molecular Weight (g/mol) 229.26
Synonym 4-Amino-1-[(2S,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone, (2S-cis)-4-Amino-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone, (+)-BCH 189, 2',3'-Dideoxy-3'-thiacytidine, 2'-Deoxy-3'-thiacytidine, 3TC, NSC 620753, cis(+)-2-Hydroxymethyl-5-(cytosin-1-yl)-1,3-oxathiolane, Lamivudine EP Impurity D
SMILES C1[C@@H](O[C@@H](S1)CO)N2C=CC(=NC2=O)N

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C8H11N3O3S/c9-5-1-2-11(8(13)10-5)6-4-15-7(3-12)14-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+/m1/s1
Chemical Name or Material ent-Lamivudine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.