missing translation for 'onlineSavingsMsg'
Learn More

ent-Indacaterol Hydrochloride, TRC

Product Code. 30391584
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391584 10 mg 10mg
30400893 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391584 Supplier Toronto Research Chemicals Supplier No. TRCI49975510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ent-Indacaterol Hydrochloride
CAS 1384188-70-8
Molecular Formula C24 H28 N2 O3 . Cl H
Quantity 10 mg
Synonym 5-[(1S)-2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-2(1H)-quinolinone Hydrochloride
InChI Formula InChI=1S/C24H28N2O3.ClH/c1-3-14-9-16-11-18(12-17(16)10-15(14)4-2)25-13-22(28)19-5-7-21(27)24-20(19)6-8-23(29)26-24;/h5-10,18,22,25,27-28H,3-4,11-13H2,1-2H3,(H,26,29);1H/t22-;/m1./s1
SMILES Cl.CCc1cc2CC(Cc2cc1CC)NC[C@@H](O)c3ccc(O)c4NC(=O)C=Cc34
IUPAC Name 5-[(1S)-2-[(5,6-diethylindan-2-yl)amino]-1-hydroxy-ethyl]-8-hydroxy-1H-quinolin-2-one;hydrochloride
Molecular Weight (g/mol) 428.95
Formula Weight 428.1867
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.