missing translation for 'onlineSavingsMsg'
Learn More

ent-Ezetimibe, TRC

Product Code. 30399135
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399135 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399135 Supplier Toronto Research Chemicals Supplier No. TRCE9750055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ent-Ezetimibe
CAS 1376614-99-1
Molecular Formula C24 H21 F2 N O3
Quantity 5 mg
Synonym 2-Azetidinone, 1-(4-fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)-, (3S,4R)-, (3S,4R)-1-(4-fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxy-propyl]-4-(4-hydroxyphenyl)azetidin-2-one, (3S,4R)-1-(4-Fluorophenyl)-3-[(R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)azetidin-2-one, ent-Ezetimibe, R,S,R-Ezetimibe (USP)
InChI Formula InChI=1S/C24H21F2NO3/c25-17-5-1-15(2-6-17)22(29)14-13-21-23(16-3-11-20(28)12-4-16)27(24(21)30)19-9-7-18(26)8-10-19/h1-12,21-23,28-29H,13-14H2/t21-,22+,23-/m0/s1
SMILES O[C@H](CC[C@H]1[C@@H](N(C1=O)c2ccc(F)cc2)c3ccc(O)cc3)c4ccc(F)cc4
IUPAC Name (3S,4R)-1-(4-fluorophenyl)-3-[(3R)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)azetidin-2-one
Molecular Weight (g/mol) 409.43
Formula Weight 409.1489
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.