missing translation for 'onlineSavingsMsg'
Learn More

Enrofloxacin, MedChemExpress

Product Code. 16478508
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16478508

Brand: Medchem Express HYB0502

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Enrofloxacin (BAY Vp 2674) is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis.

Enrofloxacin (BAY Vp 2674) is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis.

Specifications

Chemical Name or Material Enrofloxacin
Physical Form Solid
CAS 93106-60-6
Color Light Yellow
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C19H22FN3O3
For Use With (Application) COVID-19-immunoregulation
Synonym BAY Vp 2674 PD160788
Solubility Information DMSO : 10 mg/mL (27.82 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.78 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4CCN(CC)CC4)=C3)C1=O)O
Molecular Weight (g/mol) 359.39
Formula Weight 359.39
Percent Purity 99.95%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.