missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enrofloxacin-d5 HI (ethyl-d5), TRC
Specifications
Specifications
| Chemical Name or Material | Enrofloxacin D5 hydroiodide (ethyl D5) |
| CAS | 1219795-24-0 |
| Molecular Formula | C19 D5 H17 F N3 O3 . H I |
| Quantity | 1 mg |
| Synonym | Enrofloxacin D5 Hydroiodide (ethyl D5), 1-Cyclopropyl-7-[4-(ethyl-1,1,2,2,2-d5)-1-piperazinyl]-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydroiodide |
| InChI Formula | InChI=1S/C19H22FN3O3.HI/c1-2-21-5-7-22(8-6-21)17-10-16-13(9-15(17)20)18(24)14(19(25)26)11-23(16)12-3-4-12;/h9-12H,2-8H2,1H3,(H,25,26);1H/i1D3,2D2; |
| SMILES | I.[2H]C([2H])([2H])C([2H])([2H])N1CCN(CC1)c2cc3N(C=C(C(=O)O)C(=O)c3cc2F)C4CC4 |
| IUPAC Name | 1-cyclopropyl-6-fluoro-4-oxo-7-[4-(1,1,2,2,2-pentadeuterioethyl)piperazin-1-yl]quinoline-3-carboxylic acid;hydroiodide |
| Molecular Weight (g/mol) | 492.3379 |
| Formula Weight | 492.1082 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?