missing translation for 'onlineSavingsMsg'
Learn More

Enrofloxacin, ≥98%, MP Biomedicals™

Product Code. 17610927 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17610927 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17610927 Supplier MP Biomedicals Supplier No. 0219901910
  • ONLINE EXCLUSIVE PRICE
    £183.00
    £171.15 / 10g
    Save £11.85
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synonym: 1-Cyclopropyl-7-(4-Ethyl-1-Piperazinyl)-6-Fluoro-1,4-Dihydro-4-Oxo-3-Quinolonecarboxylic Acid, CFPQ, Bay Vp 2674, Baytril

Enrofloxacin it is a bactericidal agent, and is typically used as an antibiotic against Staphylococcus aureus, Escherichia coli. Salmonella spp., Klebsiella spp., and Pasteurella spp. It has also been used as a plant fungicide. Usage Statement: Unless specified otherwise, MP Biomedical™ products are for research or further manufacturing use only, not for direct human use.

Specifications

Physical Form Powder
CAS 93106-60-6
Melting Point 219°C to 221°C
Color Light Yellow
Quantity 10 g
Percent Purity ≥98%
compliance-icons compliance-icons
Product Identifier
  • Enrofloxacin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Baytril
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.