missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENPP7 Monoclonal Antibody (560514)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA561498
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
ENPP7 converts sphingomyelin to ceramide. ENPP7 also has phospholipase C activity toward palmitoyl lyso-phosphocholine. ENPP7 does not appear to have nucleotide pyrophosphatase activity.
Spécification
| ENPP7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q6UWV6 | |
| ENPP7 | |
| Mouse myeloma cell line NS0-derived recombinant human ENPP-7/Alk-SMase, Pro23-Ser439. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 560514 | |
| Unconjugated | |
| ENPP7 | |
| alkaline sphingomyelin phosphodiesterase; alkaline sphingomyelinase; Alk-SMase; ectonucleotide pyrophosphatase/phosphodiesterase 7; ectonucleotide pyrophosphatase/phosphodiesterase family member 7; E-NPP 7; ENPP7; E-NPP7; Gm254; Intestinal alkaline sphingomyelinase; NPP7; NPP-7; sphingomyelinase, intestinal alkaline; UNQ3077/PRO9912 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 339221 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu