missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENPP5 Monoclonal Antibody (500915)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524212
This item is not returnable.
View return policy
Description
In Western blots, no cross-reactivity with recombinant human (rh) ENPP-2, rhENPP-7, or recombinant mouse ENPP-7 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
Specifications
| ENPP5 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9UJA9 | |
| ENPP5 | |
| Mouse myeloma cell line NS0-derived recombinant human ENPP-5 Pro25-Ser430. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| 500915 | |
| Unconjugated | |
| ENPP5 | |
| AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 59084 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction