missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-5 Rabbit anti-Human, Clone: 14, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89909-50UL
This item is not returnable.
View return policy
Description
ENPP-5 Monoclonal specifically detects ENPP-5 in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| ENPP-5 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 59084 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 14 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, KIAA0879, NPP-5 | |
| Recombinant Human ENPP-5 Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction