missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-2/Autotaxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | ENPP-2/Autotaxin |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647728
|
Novus Biologicals
NBP2-99772-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654836
|
Novus Biologicals
NBP2-99772-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ENPP-2/Autotaxin Polyclonal antibody specifically detects ENPP-2/Autotaxin in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ENPP-2/Autotaxin | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology | |
| 0.2 um filtered solution in PBS | |
| 5168 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, Extracellular lysophospholipase D, LysoPLD, PD-IALPHA, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D | |
| Produced in rabbits immunized with purified, recombinant Human ENPP-2/Autotaxin (Accession#: AAH34961.1; Asp49-Ile863) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title