missing translation for 'onlineSavingsMsg'
Learn More

ENPP-1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18411194 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411194 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411194 Supplier Novus Biologicals Supplier No. NB600816FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

ENPP-1 Polyclonal antibody specifically detects ENPP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1
Host Species Goat
Immunogen A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5167
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.