missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENPP-1 Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-816R
This item is not returnable.
View return policy
Description
ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| ENPP-1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA | |
| P22413 | |
| ENPP1 | |
| A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 85% homology to mouse and rat. | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1 | |
| Goat | |
| 140 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5167 | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction