missing translation for 'onlineSavingsMsg'
Learn More

Enoxacin Sesquihydrate, TRC

Product Code. 30381962
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381962 100 mg 100mg
30378760 50 mg -
30383686 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381962 Supplier Toronto Research Chemicals Supplier No. TRCE557603100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84294-96-2
Molecular Formula 2 C15 H17 F N4 O3 . 3 H2 O
Molecular Weight (g/mol) 694.68
Synonym 1-Ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-1,8-Naphthyridine-3-carboxylic Acid Hydrate (2:3), Enoxacin Sesquihydrate, 1,8-Naphthyridine-3-carboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, hydrate (2:3) (9CI, ACI)
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-yl-1,8-naphthyridine-3-carboxylic acid;trihydrate
SMILES O.O.O.CCN1C=C(C(=O)O)C(=O)c2cc(F)c(nc12)N3CCNCC3.CCN4C=C(C(=O)O)C(=O)c5cc(F)c(nc45)N6CCNCC6

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C15H17FN4O3.3H2O/c2*1-2-19-8-10(15(22)23)12(21)9-7-11(16)14(18-13(9)19)20-5-3-17-4-6-20;;;/h2*7-8,17H,2-6H2,1H3,(H,22,23);3*1H2
Formula Weight 694.2886
Chemical Name or Material Enoxacin sesquihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.