missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOX2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£361.00
Specifications
| Antigen | ENOX2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
ENOX2 Polyclonal specifically detects ENOX2 in Human samples. It is validated for Western Blot.Specifications
| ENOX2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 2% Sucrose | |
| 10495 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| COVA1, Cytosolic ovarian carcinoma antigen 1APK1 antigen, ecto-NOX disulfide-thiol exchanger 2, tNOXAPK1, Tumor-associated hydroquinone oxidase | |
| The immunogen is a synthetic peptide directed towards the middle region of Human ENOX2 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title