missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ENOX2 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142286

Product Code. 17836502

  • £406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Additional Information: This antibody is expected to recognise both reported isoforms (NP_006366.2; NP_872114.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ENOX2
Polyclonal
Unconjugated
Enox2
APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197
Goat
Ammonium sulfate precipitation
RUO
10495
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin)
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q16206
Enox2
Peptide with sequence C-EKLKDDKLQVEK.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.