missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106314
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ENOX2.
This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.
Specifications
| ENOX2 | |
| Polyclonal | |
| Unconjugated | |
| Enox2 | |
| APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10495, 209224 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16206, Q8R0Z2 | |
| Enox2 | |
| A synthesized peptide derived from human ENOX2(Accession Q16206), corresponding to amino acid residues G53-C103. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction