missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENOX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545041
This item is not returnable.
View return policy
Description
Peptide sequence: YRIRLGSSTD KKDTGRLHVD FAQARDDLYE WECKQRMLAR EERHRRRMEE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.
Specifications
| ENOX2 | |
| Polyclonal | |
| Unconjugated | |
| Enox2 | |
| APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10495 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q16206 | |
| Enox2 | |
| Synthetic peptide directed towards the N-terminal of human ENOX2 (aa 184-233). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction