missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ENOX2 Polyclonal antibody specifically detects ENOX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ENOX2 fragment. |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?