Learn More
Invitrogen™ ENOX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113709
Description
Immunogen sequence: QQLQFLQQTM QGMQQQLLTI QEELNNKKSE LEQAKEEQSH TQALLKVLQE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%.
Probably acts as a terminal oxidase of plasma electron transport from cytosolic NAD(P)H via hydroquinones to acceptors at the cell surface. Hydroquinone oxidase activity alternates with a protein disulfide-thiol interchange/oxidoreductase activity which may control physical membrane displacements associated with vesicle budding or cell enlargement. The activities oscillate with a period length of 24 minutes and play a role in control of the ultradian cellular biological clock.
Specifications
| ENOX1 | |
| Polyclonal | |
| Unconjugated | |
| ENOX1 | |
| bA64J21.1; candidate growth-related and time keeping constitutive hydroquinone (NADH) oxidase; candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase; cCNOX; Cell proliferation-inducing gene 38 protein; CNOX; Constitutive Ecto-NOX; ecto-NOX disulfide-thiol exchanger 1; ENOX1; Hydroquinone [NADH] oxidase; PIG38; Protein disulfide-thiol oxidoreductase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55068 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TC92 | |
| ENOX1 | |
| Synthetic peptide directed towards the middle region of human ENOX1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.