missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOSF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ENOSF1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ENOSF1 Polyclonal specifically detects ENOSF1 in Human samples. It is validated for Western Blot.Specifications
| ENOSF1 | |
| Polyclonal | |
| Rabbit | |
| Q7L5Y1 | |
| 55556 | |
| Synthetic peptides corresponding to ENOSF1(enolase superfamily member 1) The peptide sequence was selected from the N terminal of ENOSF1. Peptide sequence MVRGRISRLSVRDVRFPTSLGGHGADAMHTDPDYSAAYVVIETDAEDGIK. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Antisense RNA to thymidylate synthase, EC 5.-, enolase superfamily member 1, HSRTSBETA, rTS, rTS beta, RTSrTS alpha, TYMSASmitochondrial enolase superfamily member 1 | |
| ENOSF1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title