missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eNOS Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-500AF350
This item is not returnable.
View return policy
Description
eNOS Polyclonal specifically detects eNOS in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Drosophila, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| eNOS | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII | |
| Rabbit | |
| 0.1 mL | |
| 4846 | |
| Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Drosophila, Sheep | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NOS3 | |
| Synthetic peptide corresponding to residues S(1179) LQERQLRGAVPWAFD(1194) of human eNOS. | |
| Primary | |
| This does not detect human inducible NOS or rat brain NOS. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction