missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ENOPH1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ENOPH1.
Bifunctional enzyme that catalyzes the enolization of 2,3-diketo-5-methylthiopentyl-1-phosphate (DK-MTP-1-P) into the intermediate 2-hydroxy-3-keto-5-methylthiopentenyl-1-phosphate (HK-MTPenyl-1-P), which is then dephosphorylated to form the acireductone 1,2-dihydroxy-3-keto-5-methylthiopentene (DHK-MTPene).
Specifications
Specifications
| Antigen | ENOPH1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ENOPH1 |
| Gene Accession No. | Q5PPH0, Q8BGB7, Q9UHY7 |
| Gene Alias | 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; 2,3-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255; 2310057D15Rik; acireductone synthase; BB183658; C81437; E1; E-1 enzyme; enolase-phosphatase 1; enolase-phosphatase E1; enolase-phosphatase E1 {ECO:0000255; ENOPH1; HAMAP-Rule:MF_03117}; Masa; MASA homolog; MASA homolog {ECO:0000255; MST145; MSTP145; mtnC; RGD1309016 |
| Gene Symbols | ENOPH1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?