missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOPH1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | ENOPH1 |
|---|---|
| Clone | 1F1E11 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234929
|
Proteintech
68179-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235225
|
Proteintech
68179-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ENOPH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ENOPH1. This antibody recognizes human, mouse, rat, rabbit, pig antigen. The ENOPH1 antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.Specifications
| ENOPH1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 58478 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1F1E11 | |
| Western Blot, ELISA, Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Rabbit, Pig | |
| DKFZp586M0524; E1; FLJ12594; MASA; MST145 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel