missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Enolase 2/Neuron-specific Enolase Antibody (NSE-P1), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Enolase 2/Neuron-specific Enolase |
|---|---|
| Clone | NSE-P1 |
| Dilution | Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30067184
|
Novus Biologicals™
NBP3-30894-200ug |
200 μg |
£386.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067154
|
Novus Biologicals™
NBP3-30894-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enolase 2/Neuron-specific Enolase Monoclonal antibody specifically detects Enolase 2/Neuron-specific Enolase in Chicken,Human,Mouse samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)Spécification
| Enolase 2/Neuron-specific Enolase | |
| Western Blot : 1-2 μg/mL, ELISA, Immunohistochemistry-Paraffin : 1-5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Chicken, Human, Mouse | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the human Enolase 2/Neuron-specific Enolase protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse and chicken. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| NSE-P1 | |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| Phosphate buffered saline | |
| 2026 | |
| IgG1 | |
| Protein A or G purified |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit