missing translation for 'onlineSavingsMsg'
Learn More

Enolase 1 Antibody (253), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

£157.00 - £382.00

Specifications

Antigen Enolase 1
Clone 253
Concentration 1 mg/mL
Dilution Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000, Immunoassay
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18163493
View Documents
Novus Biologicals
NBP2-25147SS
0.025 mL
£157.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18779983
View Documents
Novus Biologicals
NBP2-25147
0.1 mL
£382.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Enolase 1 Monoclonal specifically detects Enolase 1 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay.
Specifications

Specifications

Enolase 1
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
Unconjugated
Mouse
Cancer, Core ESC Like Genes, Stem Cell Markers
50% PBS, 50% glycerol with 5mM Sodium Azide
2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin
ENO1
IgG1
Protein G purified
Human, Bovine, Mouse, Rat, Xenopus. Other species have not been tested.
253
Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000, Immunoassay
Monoclonal
Purified
RUO
Human, Mouse, Rat, Pig, Bovine, Equine
P06733
2023
The N-terminal 12 amino acids of bovine Enolase 1, which was synthesized on a 8-amine lysine core. [UniProt# Q9XSJ4]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
47 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.