missing translation for 'onlineSavingsMsg'
Learn More

Enolase 1 Antibody (253), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18425915 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425915 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425915 Supplier Novus Biologicals Supplier No. NBP225147AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Enolase 1 Monoclonal antibody specifically detects Enolase 1 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Enolase 1
Applications Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Immunoassay
Classification Monoclonal
Clone 253
Conjugate Alexa Fluor 350
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay
Formulation 50mM Sodium Borate
Gene Alias 2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin
Host Species Mouse
Immunogen The N-terminal 12 amino acids of bovine Enolase 1, which was synthesized on a 8-amine lysine core. [UniProt# Q9XSJ4]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2023
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.