missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enolase 1 Antibody (186), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43701
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
Enolase 1 Monoclonal antibody specifically detects Enolase 1 in Human, Rat samples. It is validated for Western Blot.
Specifications
| Enolase 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| ENO1 | |
| Recombinant protein encompassing a sequence within the center region of human ENO1. The exact sequence is proprietary. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| 186 | |
| Western Blot 1:5000-1:20000 | |
| 2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin | |
| Mouse | |
| 47 kDa | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 2023 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction